Orders LinkContactUsLinkComponent
L(-)-Carnitine
L(-)-Carnitine
Glass bottle for solids, 10 g
Catalog # BMOL53922-2
Supplier:   Biomol Enzo
L(-)-Carnitine
Catalog # BMOL53922-2
Supplier:  Biomol Enzo
CAS Number:  541-15-1
L(-)-Carnitine
Glass bottle for solids, 10 g
CAS Number:  541-15-1
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Levocarnitine, L-Carnitine, Vitamin BT, (R)-3-Hydroxy-4-(trimethylammonio)butanoate, R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
  • CAS Number:541-15-1
  • Formula:C₇H₁₅NO₃
  • MDL Number:MFCD00038747
  • Molecular Weight:161.20 g/mol
  • EINECS:208-768-0
Specifications
Specifications
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    10 g