Petri dishes, PYREX®
Piatto Petri, in vetro, 80 mm, 20 mm, Non sterili
Catalog # 391-2024
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, with lid.
- Chemically resistant
- Specially formed to ensure even wall thickness and consistent optical performance
- Able to withstand repeated autoclaving
Specifications
- Ø:80 mm
- Sterilità:Non sterili
- Versione:Piatto Petri, in vetro
- Altezza:20 mm
- Seq:03