Sampling cannulae
Bent cannula with Luer lock, Acciaio inossidabile 316, 330 mm
Codice catalogo QLAACAN013-ST-9
Nome del fornitore: QUALITY LAB ACCESSORIES
Vai a
Product Details & Documents
Standard cannula are unique, proprietary configurations for most manual and automated sampling applications.
- Manufactured in both electropolished 316 stainless steel or PEEK
- Can accommodate all standard tip filters
- Optional stoppers set the proper sampling position
The USP defines the sampling point as: 'A zone half way between the top of the media and top of the paddle or basket not less than 1 cm from the vessel wall. The sampling position is critical but so too is the type of sampling cannula (also known as a probe) used.'
Specifiche
- Descrizione:Bent cannula with Luer lock
- Lunghezza:330 mm
- Materiale:Acciaio inossidabile 316
- Seq:22
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Stopper for adjustable cannula, Hanson, Ø = 3,2 mm