Boc-D-Pro-OH ≥98%
Glass bottle for solids, 5 g
Purity: 98+% CAS No.: 37784-17-1 Molecular Formula: C10H17NO4 Formula Weight: 215.25 Storage Temperature: 2°C to 8°C Melting Point: 133-135° Beilstein Reference: 479316 Optical Rotation: +61° (c=1 in acetic acid) MDL No.: MFCD00063226
: (2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline
- :37784-17-1
- :C₁₀H₁₇NO₄
- :MFCD00063226
- :215.25 g/mol
- :133…135 °C
Documents
- Tipo di conf.:Glass bottle for solids
- Conf:5 g