Orders LinkContactUsLinkComponent
Boc-D-Pro-OH ≥98%
Glass bottle for solids, 5 g
  L08826.06
Boc-D-Pro-OH ≥98%
  L08826.06
 :  37784-17-1
Boc-D-Pro-OH ≥98%
Boc-D-Pro-OH ≥98%
Glass bottle for solids, 5 g
 :  37784-17-1
SFr. 343.00
1 * 5 g
Purity: 98+% CAS No.: 37784-17-1 Molecular Formula: C10H17NO4 Formula Weight: 215.25 Storage Temperature: 2°C to 8°C Melting Point: 133-135° Beilstein Reference: 479316 Optical Rotation: +61° (c=1 in acetic acid) MDL No.: MFCD00063226
 :  (2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline
  •  :37784-17-1
  •  :C₁₀H₁₇NO₄
  •  :MFCD00063226
  •  :215.25 g/mol
  •  :133…135 °C
  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    5 g