Orders LinkContactUsLinkComponent
Bond Elut adapters
Bond Elut adapters
Adapter cap for 60 ml Bond Elut cartridges
Catalog # VARI12131004
Supplier:   VARIAN
Bond Elut adapters
Catalog # VARI12131004
Supplier:  VARIAN
Bond Elut adapters
Adapter cap for 60 ml Bond Elut cartridges
Price
Inquire For Price
Log in for availability
1 * 10 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Bond Elut adapters fit on top of any Bond Elut cartridge and contain a female Luer fitting that accommodates the tip of another cartridge.
  • Connect SPE cartridges in series for large samples
  • Expand cartridge volume for even more applications
  • Transfer large-volume samples to any SPE cartridge
Specifications
  • Descrizione:
    Adapter cap for 60 ml Bond Elut cartridges
  • Seq:
    02