Orders LinkContactUsLinkComponent
First-aid kits
First-aid kits
Kit grande, 21 - 50 persone
Codice catalogo 118-0130
Nome del fornitore:   SAFETY FIRST AID GROUP
First-aid kits
Codice catalogo 118-0130
Nome del fornitore:  SAFETY FIRST AID GROUP
First-aid kits
First-aid kits
Kit grande, 21 - 50 persone
Price
165.00 SFr.
Log in per disponibilità
1 * 1 KIT
Product Details & Documents
The contents of the following first aid kits meet the requirements as specified in the HSE Approved Code of Practice. The code of practice states 'An employer should make an assessment of the first aid needs appropriate to the circumstance of each workplace'. To assist in selecting the appropriate kit, employee ratios are given as guidance provided by the British Healthcare Trade Association.
Informazioni per l'ordine: All kits are supplied with a first aid guidance leaflet.
Specifiche
  • Descrizione:
    Kit grande, 21 - 50 persone
  • Cat. No.:
    118-0130
  • Contenuti:
    60 × Waterproof plasters, sterile
    20 × individually wrapped wipes
    6 × eye pads with bandages, non woven
    8 × triangular bandages, non woven
    12 × Safety pins
    12 × sterile dressing, medium size
    4 × sterile dressing, large size
    3 pairs of disposable examination gloves
    1 wall bracket
  • Modello:
    Standard (21 - 50 persons)