Combustion boats
103×20×12 mm
Codice catalogo TSLA73382
Nome del fornitore: TECHNICAL QUARTZ SOLUTIONS
Vai a
Product Details & Documents
Combustion boats, quartz, glazed, with or without handle.
- Very good chemical resistance
- Very good temperature resistance
Specifiche
- Seq:02
- L×P×A:103×20×12 mm