Orders LinkContactUsLinkComponent
Cleanroom squeegee tool, NovaSqueegee™
Cleanroom squeegee tool, NovaSqueegee™
Lama, in gomma per adattatore 48,72 cm (18")
Catalog # 141-0097
Supplier:   MICRONOVA
Cleanroom squeegee tool, NovaSqueegee™
Catalog # 141-0097
Supplier:  MICRONOVA
Cleanroom squeegee tool, NovaSqueegee™
Cleanroom squeegee tool, NovaSqueegee™
Lama, in gomma per adattatore 48,72 cm (18")
Price
SFr. 21.20
Log in for availability
1 * 1 ST
Product Details & Documents
A stainless steel squeegee tool with a replaceable rubber blade, designed to clean windows, screens, floors, and walls in the cleanroom. Optional PVA blades are fast-drying, low-linting, and low in particulates with a 600% absorbency rate; ideal for electronic and semiconductor cleanrooms.
  • Designed to clean windows, screens, floors, and walls in the cleanroom
  • Made with 304 electropolished stainless steel and Derlin® resin
  • Rubber replacement blades available
  • PVA replacement blades available for absorbency
  • Quick disconnect handle attachment
  • Entire unit is autoclavable
The Micronova Cleanroom NovaSqueegee™ is a stainless steel tool that comes standard with a replaceable rubber blade and features a quick disconnect handle attachment for optional use with the SSU series handles for extended reach.
Specifications
  • Descrizione:
    Lama, in gomma per adattatore 48,72 cm (18")
  • Seq:
    07