Orders LinkContactUsLinkComponent
Cleanroom squeegee tool, NovaSqueegee™
Cleanroom squeegee tool, NovaSqueegee™
Adattatore 22'' - 56 cm
Codice catalogo 129-0533
Nome del fornitore:   MICRONOVA
Cleanroom squeegee tool, NovaSqueegee™
Codice catalogo 129-0533
Nome del fornitore:  MICRONOVA
Cleanroom squeegee tool, NovaSqueegee™
Cleanroom squeegee tool, NovaSqueegee™
Adattatore 22'' - 56 cm
Price
SFr. 265.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
A stainless steel squeegee tool with a replaceable rubber blade, designed to clean windows, screens, floors, and walls in the cleanroom. Optional PVA blades are fast-drying, low-linting, and low in particulates with a 600% absorbency rate; ideal for electronic and semiconductor cleanrooms.
  • Designed to clean windows, screens, floors, and walls in the cleanroom
  • Made with 304 electropolished stainless steel and Derlin® resin
  • Rubber replacement blades available
  • PVA replacement blades available for absorbency
  • Quick disconnect handle attachment
  • Entire unit is autoclavable
The Micronova Cleanroom NovaSqueegee™ is a stainless steel tool that comes standard with a replaceable rubber blade and features a quick disconnect handle attachment for optional use with the SSU series handles for extended reach.
Specifiche
  • Descrizione:
    Adattatore 22'' - 56 cm
  • Seq:
    03