Orders LinkContactUsLinkComponent
Cleanroom squeegee tool, NovaSqueegee™
Adattatore 18'' - 46 cm
  129-0532
 :   MICRONOVA
Cleanroom squeegee tool, NovaSqueegee™
  129-0532
 :  MICRONOVA
Cleanroom squeegee tool, NovaSqueegee™
Cleanroom squeegee tool, NovaSqueegee™
Adattatore 18'' - 46 cm
SFr. 260.00
1 * 1 ST
A stainless steel squeegee tool with a replaceable rubber blade, designed to clean windows, screens, floors, and walls in the cleanroom. Optional PVA blades are fast-drying, low-linting, and low in particulates with a 600% absorbency rate; ideal for electronic and semiconductor cleanrooms.
  • Designed to clean windows, screens, floors, and walls in the cleanroom
  • Made with 304 electropolished stainless steel and Derlin® resin
  • Rubber replacement blades available
  • PVA replacement blades available for absorbency
  • Quick disconnect handle attachment
  • Entire unit is autoclavable
The Micronova Cleanroom NovaSqueegee™ is a stainless steel tool that comes standard with a replaceable rubber blade and features a quick disconnect handle attachment for optional use with the SSU series handles for extended reach.
  • Descrizione:
    Adattatore 18'' - 46 cm
  • Seq:
    02