Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Conductive grid tapes, CGT™

Conductive grid tapes, CGT™

Nastro conduttivo a griglia CGT™ per camera bianca, -, 50,8 mm
Codice catalogo 819-0076
Nome del fornitore:   MICRONOVA
Conductive grid tapes, CGT™
Codice catalogo 819-0076
Nome del fornitore:  MICRONOVA
Conductive grid tapes, CGT™
Conductive grid tapes, CGT™

Nastro conduttivo a griglia CGT™ per camera bianca, -, 50,8 mm

Price
SFr. 67.40
Log in per disponibilità
1 * 1 ROU
Product Details & Documents

Conductive, anti-static polypropylene tape, specially designed with conductive grids embedded between two anti-static layers, for use on devices in ESD sensitive environments.

  • Heavy-duty, anti-static copolymer tape with acrylic adhesive
  • Manufactured for use in ESD sensitive environments
  • Features a conductive grid embedded between two antistatic layers
  • Conducts charge away from substrate
  • Protects sensitive devices that would be otherwise damaged by normal insulating tape
  • Provides an effective static shield and prevents static buildup
  • Prevents shedding, flaking, cracking, or chipping of the grids
  • 36 yards wound on a 3" plastic core

The Micronova CGT™ Conductive Grid Tape features conductive grids that are embedded between two anti-static layers to provide an effective static shield. This design prevents static build-up, as well as preventing any shedding, flaking, cracking, or chipping of the grids.

 : For additional widths, contact your Avantor sales representative.

Specifiche
  • Sterili:
    -
  • Larghezza:
    50,8 mm
  • Descrizione:
    Nastro conduttivo a griglia CGT™ per camera bianca
  • Seq:
    60
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.