Surgical scissors, type Mayo
Curved, bevelled blades, blunt, 165 mm
Codice catalogo MURR1882/10
Nome del fornitore: MURRAY S
Vai a
Product Details & Documents
These scissors can be used for a variety of applications in medical areas, laboratories and microscopy.
- Straight or curved
- Bevelled blades
- Blunt
Specifiche
- Tipo:Curved, bevelled blades, blunt
- Lunghezza:165 mm
- Seq:03