Sponge swabs, Polywipe®
Polywipe, confezione con chiusura a strappo
720-0145
Non-inhibitory sponge material allows effective sampling of larger areas for environmental and hygiene monitoring. Available dry or pre-moistened with sterile diluent. Polywipes are supplied in peel pouches for immediate use, or in a 110 ml blue-capped container for transfer to the laboratory.
- Blue colour for visibility
- Triple wrapped and radiation sterilised
Delivery information: Supplied with individual sterile glove for handling.
- Descrizione:Polywipe, confezione con chiusura a strappo