ISE sample conditioning buffers and solutions
ISA/Na, ionic strength adjustment solution for sodium, 11, Bottle, 1 L
Nome del fornitore: WTW
ISE sample conditioning buffers and solutions
Codice catalogo 662-0379
Nome del fornitore: WTW
Numero CAS:
Vai a
Product Details & Documents
Ion strength adjustment solutions have multiple tasks: TISAB for fluoride measurements does not only adjust the pH value, by also decomplexes the fluoride-containing complex compounds. Other ion strength adjustment solutions ensure an adapted pH value as well as the addition of a suitable conducting salt to correctly capture the concentration.
Documents
Specifiche
Specifiche
- Tipo di conf.:Bottle
- Conf:1 L
- Descrizione:ISA/Na, ionic strength adjustment solution for sodium
- pH:11