Orders LinkContactUsLinkComponent
Stirrer blades, PTFE, Quickfit®
Stirrer blades, PTFE, Quickfit®
Stirrer blade, with 19/26 socket to fit ST6/7, 52 mm
Codice catalogo 441-0359
Nome del fornitore:   SciLabware
Stirrer blades, PTFE, Quickfit®
Codice catalogo 441-0359
Nome del fornitore:  SciLabware
Stirrer blades, PTFE, Quickfit®
Stirrer blades, PTFE, Quickfit®
Stirrer blade, with 19/26 socket to fit ST6/7, 52 mm
Price
SFr. 64.20
Log in per disponibilità
1 * 5 ST
Product Details & Documents
Stirrer blades made of PTFE, and they can be pivoted on the glass stirrer shaft to allow passage through small apertures.
  • Easily detachable for cleaning and replacement purposes
  • Resistance to chemical attack
Suitable for sweeping close to the vessel walls as they will not cause scratching.
Specifiche
  • Tipo:
    Stirrer blade, with 19/26 socket to fit ST6/7
  • Larghezza:
    52 mm
  • Seq:
    01