Stirrer blades, PTFE, Quickfit®
Stirrer blade, with 19/26 socket to fit ST6/7, 52 mm
Codice catalogo 441-0359
Nome del fornitore: SciLabware
Vai a
Product Details & Documents
Stirrer blades made of PTFE, and they can be pivoted on the glass stirrer shaft to allow passage through small apertures.
- Easily detachable for cleaning and replacement purposes
- Resistance to chemical attack
Suitable for sweeping close to the vessel walls as they will not cause scratching.
Specifiche
- Tipo:Stirrer blade, with 19/26 socket to fit ST6/7
- Larghezza:52 mm
- Seq:01