Filtration funnels, Quickfit®
P40, 19/26 NS, 30 mm
Catalog # 201-8685
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered glass disc.
Cone connection.
Specifications
- Dimensione cono:19/26 NS
- Ø piastra:30 mm
- Porosità:P40