Dressing jars
100 mm, 100 mm
Codice catalogo MURRG500/01
Nome del fornitore: MURRAY S
Vai a
Product Details & Documents
Dressing jars, made of moulded glass, round.
- Good chemical resistance
- Lids must be ordered separately
Specifiche
- Ø est:100 mm
- Altezza:100 mm
- Seq:01