Burettes, removable stopcock
Glass jet for 25 and 50 ml burettes
Codice catalogo BIBBBRB284
Nome del fornitore: SciLabware
Vai a
Product Details & Documents
Soda lime glass, class B, removable PTFE stopcock.
- Flared top for safer filling
- Blue enamel graduations
BS 846
Certifications: BS 846
Documents
Specifiche
- Descrizione:Glass jet for 25 and 50 ml burettes
- Seq:05