Polysorbate 60
bottiglia in vetro, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 9005-67-8 Molecular Formula: C64H126O26 Flash Point: 150°(302°F) Merck Reference: 14,7582 MDL No.: MFCD01780110
Synonyms: Sorbitan monostearate, ethoxylated, Polyoxyethylene sorbitan monostearate
- CAS Number:9005-67-8
- Formula:C₆₄H₁₂₆O₂₆
- MDL Number:MFCD01780110
- Merck Index:14,7582
- Density:1,080
- Flash Pt:>150 °C (302 °F)
- Storage Temperature:Ambiente
- EINECS:500-020-4
Specifications
Specifications
- Tipo di conf.:bottiglia in vetro
- Conf:100 mL



