Orders LinkContactUsLinkComponent
Accessories for comparators | Avantor
Accessories for comparators
Chlorine, 3/2APH, 2,0-10,0 mg/l
Catalog # LOVI232060
Supplier:   Lovibond
Accessories for comparators
Catalog # LOVI232060
Supplier:  Lovibond
Accessories for comparators
Accessories for comparators
Chlorine, 3/2APH, 2,0-10,0 mg/l
Price
299.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+
Specifications
  • Range di misura:
    2,0-10,0 mg/l
  • Seq:
    18
  • Codice:
    3/2APH
  • Metodo di prova:
    Chlorine
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..