Accessories for comparators
QAC (Quaternary Ammonia Compounds), 3/118, 2 - 20 mg/l
Codice catalogo LOVI230120
Nome del fornitore: Lovibond
Vai a
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
- Disc codes starting with 'N' are for use with the Nessleriser 2150
- Disc codes starting with 'C' are for use with the Nessleriser 2250
- All other discs are for use with the Comparator 2000+
Specifiche
- Range di misura:2 - 20 mg/l
- Seq:71
- Codice:3/118
- Metodo di prova:QAC (Quaternary Ammonia Compounds)