Orders LinkContactUsLinkComponent
Stoppers, with drip tip, MBL®
19/26, With pointed bottom
Codice catalogo 217-0747
Nome del fornitore:   SciLabware
Stoppers, with drip tip, MBL®
Codice catalogo 217-0747
Nome del fornitore:  SciLabware
Stoppers, with drip tip, MBL®
Stoppers, with drip tip, MBL®
19/26, With pointed bottom
Price
98.50 SFr.
Log in per disponibilità
1 * 10 ST
Product Details & Documents
Ideally suited for use with volumetric flasks etc. Lower closed end of stopper features a 'drip-tip' to ensure adhered liquid is returned to the flask. Flat head allows stopper to be stood on end, thus preventing contamination of the ground surface.
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Hollow-blown to reduce weight
Specifiche
  • Cono (NS):
    19/26
  • Tipo:
    With pointed bottom
  • Seq:
    05