Burettes, removable stopcock
Glass burette tube, 50 ml
Codice catalogo BIBBBRB276P
Nome del fornitore: SciLabware
Vai a
Product Details & Documents
Soda lime glass, class B, removable PTFE stopcock.
- Flared top for safer filling
- Blue enamel graduations
BS 846
Certifications: BS 846
Documents
Specifiche
- Descrizione:Glass burette tube, 50 ml
- Seq:04