Orders LinkContactUsLinkComponent
Büchner funnels, Quickfit®
Büchner funnels, Quickfit®
55 mm, 3, 24/29 NS
Codice catalogo 532-0097
Nome del fornitore:   SciLabware
Büchner funnels, Quickfit®
Codice catalogo 532-0097
Nome del fornitore:  SciLabware
Büchner funnels, Quickfit®
Büchner funnels, Quickfit®
55 mm, 3, 24/29 NS
Price
141.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered glass disc and integral ground glass cone.
Specifiche
  • Dimensione cono:
    24/29 NS
  • Ø piastra:
    55 mm
  • Porosità:
    3