Orders LinkContactUsLinkComponent
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
100 ml, 14/23 NS, 14/23 NS
Codice catalogo 532-0082
Nome del fornitore:   SciLabware
Separating funnels, pear shaped, Quickfit®
Codice catalogo 532-0082
Nome del fornitore:  SciLabware
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
100 ml, 14/23 NS, 14/23 NS
Price
153.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Excellent chemical resistance
  • Excellent temperature resistance
Funnel stem with cone to allow easy connection to Quickfit® flask with interchangeable PTFE key stopcock.
Certifications: Complies with ISO 4800 type 2.
Specifiche
  • Dimensione cono:
    14/23 NS
  • Cono femmina:
    14/23 NS
  • Capacità:
    100 ml
  • Seq:
    04