Orders LinkContactUsLinkComponent
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
50 ml, 14/23 NS, 14/23 NS
Catalog # 532-0080
Supplier:   SciLabware
Separating funnels, pear shaped, Quickfit®
Catalog # 532-0080
Supplier:  SciLabware
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
50 ml, 14/23 NS, 14/23 NS
Price
SFr. 153.00
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Excellent chemical resistance
  • Excellent temperature resistance
Funnel stem with cone to allow easy connection to Quickfit® flask with interchangeable PTFE key stopcock.
Certifications: Complies with ISO 4800 type 2.
Specifications
  • Dimensione cono:
    14/23 NS
  • Cono femmina:
    14/23 NS
  • Capacità:
    50 ml
  • Seq:
    02