Orders LinkContactUsLinkComponent
Dropping funnels, cylindrical, Quickfit®
Dropping funnels, cylindrical, Quickfit®
100 ml, 19/26 NS, 19/26 NS
Codice catalogo 532-0076
Nome del fornitore:   SciLabware
Dropping funnels, cylindrical, Quickfit®
Codice catalogo 532-0076
Nome del fornitore:  SciLabware
Dropping funnels, cylindrical, Quickfit®
Dropping funnels, cylindrical, Quickfit®
100 ml, 19/26 NS, 19/26 NS
Price
114.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Funnel stem with ground-glass cone for flasks and assemblies. Stopcock has interchangeable glass key.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Certifications: Complies with ISO 4800
Specifiche
  • Dimensione cono:
    19/26 NS
  • Cono femmina:
    19/26 NS
  • Capacità:
    100 ml
  • Seq:
    02