Powder funnels, Quickfit®
34/35 NS, 105 mm
Catalog # 532-0073
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, stem with cone.
- Ottima resistenza ai prodotti chimici
- Resistenza alle alte temperature
Designed for transfer of powders into flasks and vessels.
Specifications
- Dimensione cono:34/35 NS
- Ø esterno imbuto:105 mm
- Seq:05