Counting chambers, Thoma
Counting chamber, bright lines, Thoma
Codice catalogo 720-1899
Nome del fornitore: MARIENFELD
Vai a
Product Details & Documents
Designed for the determination of the yeast cell number and IVD applications.
Certifications: IVD directive 98/79/EC.
Specifiche
- Descrizione:Counting chamber, bright lines, Thoma
- Seq:3