Orders LinkContactUsLinkComponent
Weighing boats, rounded
Weighing boats, rounded
1,5 ml, 70 mm
Codice catalogo NEUB1261-17207
Nome del fornitore:   NEUBERT VOLUME GLASSWAERE
Weighing boats, rounded
Codice catalogo NEUB1261-17207
Nome del fornitore:  NEUBERT VOLUME GLASSWAERE
Weighing boats, rounded
1,5 ml, 70 mm
Price
SFr. 60.20
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Weighing funnels, borosilicate glass 3.3, designed to weigh chemicals or for weighing chemicals with tubular stalk.
  • Excellent chemical resistance
  • Excellent temperature Resistance
Specifiche
  • Capacità:
    1,5 ml
  • Lunghezza:
    70 mm
  • Seq:
    02