Orders LinkContactUsLinkComponent
Counting chambers, Thoma
Counting chambers, Thoma
Counting chamber, dark lines, Thoma
Codice catalogo 631-0697
Nome del fornitore:   MARIENFELD
Counting chambers, Thoma
Codice catalogo 631-0697
Nome del fornitore:  MARIENFELD
Counting chambers, Thoma
Counting chambers, Thoma
Counting chamber, dark lines, Thoma
Price
62.80 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Designed for the determination of the yeast cell number and IVD applications.
Certifications: IVD directive 98/79/EC.
Specifiche
  • Descrizione:
    Counting chamber, dark lines, Thoma
  • Seq:
    2