Counting chambers, Malassez
Cell counting chamber with dark lines, Malassez, 0,2 mm
Nome del fornitore: MARIENFELD
Vai a
Product Details & Documents
Counting chambers specially designed for in vitro diagnostic.
- Double net ruling
- Two cover glasses
- Dark lined without clamps
Documents
Specifiche
- Profondità della camera:0,2 mm
- Tipo:Malassez
- Descrizione:Cell counting chamber with dark lines
- Seq:001