Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Provette per centrifuga

Provette per centrifuga

15 ml, ∼17×115 mm, -, graduated
Codice catalogo HIRS6020101
Nome del fornitore:   HIRSCHMANN
Provette per centrifuga
Codice catalogo HIRS6020101
Nome del fornitore:  HIRSCHMANN
Provette per centrifuga
Provette per centrifuga

15 ml, ∼17×115 mm, -, graduated

Price
SFr. 412.00
Log in per disponibilità
1 * 100 ST
Product Details & Documents

Glass.

  • Graduated or ungraduated
  • Good chemical resistance

Long conical or short conical form.

Specifiche
  • Tipo di chiusura:
    -
  • Ø×L:
    ∼17×115 mm
  • Tipo:
    graduated
  • Capacità:
    15 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.