Orders LinkContactUsLinkComponent
Conical cork stoppers
30,0 mm, 70,0 mm, 75,0 mm
  BRAMKS30-75-70
 :   BRAEMSWIG
Conical cork stoppers
  BRAMKS30-75-70
 :  BRAEMSWIG
Conical cork stoppers
30,0 mm, 70,0 mm, 75,0 mm
SFr. 79.70
1 * 10 ST
These conical stoppers are made of cork.
  • Ø inferiore:
    70,0 mm
  • Ø superiore:
    75,0 mm
  • Altezza:
    30,0 mm
  • Seq:
    13