Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

1,3-Propansultone, semi-solid

Glass bottle for solids, 10 g
Catalog # ICNA0215639110
Health
Irritant
Supplier:   MP Biomedicals
1,3-Propansultone, semi-solid
Catalog # ICNA0215639110
Supplier:  MP Biomedicals
CAS Number:  1120-71-4
1,3-Propansultone, semi-solid

Glass bottle for solids, 10 g

CAS Number:  1120-71-4
Price
SFr. 54.30
Log in for availability
1 * 10 g
Product Details & Documents

1,3-Propane Sultone was used as quaternisation agent to transit the PDMAEMA into poly(zwitterions) through surface reaction by treating membranes under a mild condition.

1,3-Propane Sultone was used as quaternisation agent to transit the PDMAEMA into poly(zwitterions) through surface reaction by treating membranes under a mild condition.

Soluble in cold water, hot water; readily in ketones, esters and aromatic hydrocarbons; insoluble in aliphatic hydrocarbons.

Synonyms:  1,2-Oxathiolane 2,2-dioxide
  • CAS Number:1120-71-4
  • Formula:C₃H₆O₃S
  • MDL Number:MFCD00005355
  • Molecular Weight:122.14 g/mol
  • Boiling Pt:298 °C (1013 hPa)
  • Melting Pt:30…33 °C (lett.)
  • Density:1,392 g/ml
  • Storage Temperature:Refrigerata
  • EINECS:214-317-9
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Tipo di conf.:
    Glass bottle for solids
  • Conf:
    10 g

Specifications