Split/splitless inlet seals
Optional split/splitless weldment
Codice catalogo HEWLG1544-60585
Nome del fornitore: AGILENT HEWLETT PACKARD
Split/splitless inlet seals
Codice catalogo HEWLG1544-60585
Nome del fornitore: AGILENT HEWLETT PACKARD
Vai a
Product Details & Documents
These inlet seals have been developed to ensure that you have a consistent and inert surface to properly seal the inlet and prevent sample degradation.
- Very reproducible smooth surface, eliminating traces of machining grooves that can be the source of minute leaks
- High quality, leak-free seal
Informazioni per l'ordine: An inlet washer is provided with each inlet seal.
Specifiche
- Descrizione:Optional split/splitless weldment
- Seq:10