Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
ICP-MS tuning solutions

ICP-MS tuning solutions

Tuning solution, 6 elements in diluted nitric acid, Li, Mg, Y, Ce, Tl, Co, 1 μg/L, 2% HNO₃, 2 × 500 ml, 1
Codice catalogo HEWL5185-5959
Nome del fornitore:   AGILENT HEWLETT PACKARD
ICP-MS tuning solutions
Codice catalogo HEWL5185-5959
Nome del fornitore:  AGILENT HEWLETT PACKARD
Numero CAS:  
ICP-MS tuning solutions
ICP-MS tuning solutions

Tuning solution, 6 elements in diluted nitric acid, Li, Mg, Y, Ce, Tl, Co, 1 μg/L, 2% HNO₃, 2 × 500 ml, 1

Numero CAS:  
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents

PA Tuning solution set includes Tuning 1 and Tuning 2 solutions.

Tuning 1: 20 ppm each of Zn, Be, Cd, As; 10 ppm each of Ni, Pb, Mg; 5 ppm each of Tl, Na, Al, U, Cu, Th, Ba, Co, Sr, V, Cr, Mn, 6Li, Sc, In, Lu, Bi; 2,5 ppm each of Y, Yb; matrix 2 to 5% HNO₃
Tuning 2: 10 ppm each of Mo, Sb, Sn, Ge, Ru, Pd; 5 ppm of Ti, Ir; matrix 10% HCl and 1% HNO₃ with trace amounts of HF.

Documents

Specifiche

Specifiche

  • Concentrazione:
    1 μg/L
  • Matrice:
    2% HNO₃
  • Conf:
    1
  • Descrizione:
    Tuning solution, 6 elements in diluted nitric acid, Li, Mg, Y, Ce, Tl, Co
  • Pk Info:
    2 × 500 ml

Specifiche