Stopcocks, single-way, DURAN®
8 mm, 24 NS
SCOT286407606
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
- Replaceable NS keys
- Screw thread retaining device
Straight bore.
- Ø foro:8 mm
- Seq:03
- Rubinetto:24 NS