Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Graduated pipettes, Class B, partial delivery, DURAN®

0,2 ml, 0,001 ml, ±0,0045 ml, 3 × blu, -
  612-5251
Graduated pipettes, Class B, partial delivery, DURAN®
  612-5251
Graduated pipettes, Class B, partial delivery, DURAN®
Graduated pipettes, Class B, partial delivery, DURAN®

0,2 ml, 0,001 ml, ±0,0045 ml, 3 × blu, -

 
SFr. 163.00
   
1 * 12 ST
 

Soda-lime glass (AR-Glas®).

  • For cotton plugs
  • Marks and inscription in brown
  • Numbering: Zero at top
  • Partial delivery (type 1)
  • Calibration is based on the poured out volume (Ex)

Complies to DIN ISO 835.

Colour code complies to DIN 12621.

 : ISO 835.

 
  • Codice colore:
    3 × blu
  • Capacità:
    0,2 ml
  • oltre a DIN:
    -
  • Divisione:
    0,001 ml
  • Tolleranza:
    ±0,0045 ml