Waste Bins
Waste disposal bin, without lid, antracite, HDPE, 60 L, 400×700 mm
Catalog # SEMA2093
Supplier: SEMADENI
Jump to:
Product Details & Documents
Made of HDPE.
- Stable and are stackable
- Odourless and hermetic seal for the transport and disposal
- Adhesive sealing and triple lid seal
Round containers are suitable for safe and clean disposal of clinical, bacteriological or chemical and hazardous waste.
Certifications: UN 1H2/Y11/S (30 litre bins), max. loading 11 kg
UN 1H2/Y22/S (60 litre bins), max. loading 22 kg
Ordering information: Lids must be ordered separately.
Specifications
- Ø×A:400×700 mm
- Capacità:60 L
- Colore:antracite
- Descrizione:Waste disposal bin, without lid
- Materiale:HDPE
- Seq:02
Accessories
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Product Information
- Catalog #DescrizioneAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



