Gramine
Glass bottle for solids, 100 mg
LATOL6088-100
: LATOXAN
: 1-(1H-Indol-3-yl)-N,N-dimethylmethanamine, 3-(Dimethylaminomethyl)-Indole
- :87-52-5
- :C₁₁H₁₄N₂
- :MFCD00005629
- :174.25 g/mol
- :201-749-8
- Tipo di conf.:Glass bottle for solids
- Conf:100 mg