Software for data loggers, elproLOG ANALYZE
elproLOG ANALYZE software, full version
Catalog # ELPR800397
Supplier: ELPRO-BUCHS
Jump to:
Product Details & Documents
elproLOG ANALYZE is a software for all elpro data loggers. It provides all functions such as data analysis, reporting and configuration of data loggers.
- User friendly
- Displays statistics on measurement data
- Optional audit trail
- Multiple zoom function
Specifications
- Descrizione:elproLOG ANALYZE software, full version
- Seq:01