Buckets, round, Azlon®
500 ml, nero, 10,5 L
Codice catalogo 216-2805
Nome del fornitore: SciLabware
Vai a
Product Details & Documents
HDPE, white or black.
- Spouted buckets for easy pouring
- Handle grip for easy lifting and carrying
- Black 10 litre bucket has metal handle for extra strength
Specifiche
- Capacità:10,5 L
- Colore:nero
- Divisione:500 ml