Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Funnels with short stem

130 mm, 35 mm, 345 mm
Codice catalogo SEMA0201
Nome del fornitore:   SEMADENI
Funnels with short stem
Codice catalogo SEMA0201
Nome del fornitore:  SEMADENI
Funnels with short stem
Funnels with short stem

130 mm, 35 mm, 345 mm

Price
SFr. 25.10
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Funnels, made of HDPE, with smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.

  • Easy to clean
  • Good chemical resistance
  • Good temperature resistance
Specifiche
  • Ø esterno imbuto:
    345 mm
  • Ø esterno stelo:
    35 mm
  • Lunghezza asta:
    130 mm