Orders LinkContactUsLinkComponent
Combined pH electrode, InLab® Flow
Combined pH electrode, InLab® Flow
Cella flow-through per elettrodo InLab® Flow, termostatabile, volume di misurazione 0,5 ml, S7
Catalog # METT59904354
Supplier:   Mettler - Toledo
Combined pH electrode, InLab® Flow
Catalog # METT59904354
Supplier:  Mettler - Toledo
Combined pH electrode, InLab® Flow
Combined pH electrode, InLab® Flow
Cella flow-through per elettrodo InLab® Flow, termostatabile, volume di misurazione 0,5 ml, S7
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
A glass-body, combination pH electrode with SteadyForce™ and S7 screw head for inline measurements in flow-through cell 611.
  • SteadyForce™ reference system: Pressurised (3 bar) electrolyte ensures electrolyte flow even in viscous samples and guarantees highly reproducible results
  • Double ceramic junction: Ideal for general applications
  • Glass body is highly chemical resistant
Ordering information: Cable and flow-through cell need to be ordered separately.
Specifications
  • Connettore:
    S7
  • Descrizione:
    Cella flow-through per elettrodo InLab® Flow, termostatabile, volume di misurazione 0,5 ml
Accessories
Product Information
  • Catalog #
    Descrizione
    Lunghezza
    Availability
    Price
  • 661-0257
    Cavo InLab® S7-BNC 1,2 m
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 78.00
Product Information
  • Catalog #
    Descrizione
    Lunghezza
    Availability
    Price
  • 661-0264
    Cavo InLab® S7-4mm 1,2 m
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 86.00
Product Information
  • Catalog #
    Descrizione
    Lunghezza
    Availability
    Price
  • 661-0261
    Cavo InLab® S7-DIN 1,2 m
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 86.70
Product Information
  • Catalog #
    Descrizione
    Lunghezza
    Availability
    Price
  • 661-0265
    Cavo InLab® S7-2 mm 1,2 m
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 86.00
Product Information
  • Catalog #
    Descrizione
    Lunghezza
    Availability
    Price
  • 661-0267
    Cavo InLab® S7-Lemo 00 1,2 m
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 211.00
Product Information
  • Catalog #
    Descrizione
    Lunghezza
    Availability
    Price
  • 661-0266
    Cavo InLab® S7-RCA 1,2 m
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 84.00
Product Information
  • Catalog #
    Descrizione
    Lunghezza
    Availability
    Price
  • 661-0260
    Cavo InLab® S7-BNC IP67 1,2 m
    1,2 m
    Log in for availability
    1 * 1 ST - SFr. 86.00