Accessories for comparators
AF332 EP colour scale, 4/64E, GY1 - GY7
Catalog # TINT246450
Supplier: Lovibond Tintometer
Jump to:
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
- Disc codes starting with 'N' are for use with the Nessleriser 2150
- Disc codes starting with 'C' are for use with the Nessleriser 2250
- All other discs are for use with the Comparator 2000+
Specifications
- Range di misura:GY1 - GY7
- Seq:87
- Codice:4/64E
- Metodo di prova:AF332 EP colour scale