Orders LinkContactUsLinkComponent
Instrument trays
Instrument trays
100×200×50 mm
Codice catalogo HECH42910003
Nome del fornitore:   GLASWARENFABRIK KARL HECHT
Instrument trays
Codice catalogo HECH42910003
Nome del fornitore:  GLASWARENFABRIK KARL HECHT
Instrument trays
Instrument trays
100×200×50 mm
Price
76.40 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Glass with chrome-plated knobbed lid.
  • Can be sterilised in hot air up to 180 °C
  • Robust
  • Good chemical resistance
Multipurpose dishes with vertical walls for a variety of applications in the laboratory, field use and industry.
Specifiche
  • L×P×A:
    100×200×50 mm