Jump to:
Product Details & Documents
Synonyms: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
- CAS Number:60-00-4
- Formula:C₁₀H₁₆N₂O₈
- MDL Number:MFCD00003541
- Molecular Weight:292.25 g/mol
- Merck Index:13,03546
- Boiling Pt:614 °C (1013 hPa)
- Melting Pt:220 °C
- Density:0,86 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Flash Pt:>100 °C
- Storage Temperature:Ambiente
- EINECS:200-449-4
- REACH:01-2119486399-18
Documents
Specifications
Specification Test Results
- Solubility:0.5 g/l (20 °C)
- Melting Point:220 °C (decomposition)
- Molar Mass:292.24 g/mol
- Bulk Density:600 kg/m3
- Vapor Pressure:- 0.013 hPa (20 °C)
- Flash Point:100 °C DIN 51758
- Density:0.86 g/cm3 (20 °C)
- pH:2.5 (10 g/l, H2O, 23 °C) (slurry)
- Ignition Point:200 °C
Specifications
- Tipo di conf.:Bottiglia di plastica
- Conf:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



