Pharmacist's spatulas
Hardened blade, 130 mm, 18/22 mm
231-0553
: RSG
Stainless steel blade, wooden handle.
- Available with flexible blade (Remanit 4310) or with hardened blade (INOX 4034)
- 75 to 150 mm: conical blade
- Descrizione:Hardened blade
- Larghezza lama:18/22 mm
- Lunghezza lama:130 mm