Instrument trays, rectangular
30 mm, 180 mm, 260 mm
Codice catalogo RSGA701.226
Nome del fornitore: RSG
Vai a
Product Details & Documents
Stainless steel, 18/10.
- Smooth surfaces for fast, easy cleaning
- Good chemical resistance
- Very good thermal resistance
Documents
RSGA701.242_TDS_EN
Scheda dati
Specifiche
- Larghezza:180 mm
- Altezza:30 mm
- Lunghezza:260 mm